missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Liver Digest Medium
GREENER_CHOICE

Product Code. 11550536
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. Quantity unitSize
11550536 500 mL 500mL
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11550536

Brand: Gibco™ 17703034

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Liver Digest Medium

Liver Digest Medium is a collagenase-dispase medium for dissociation of viable liver cells.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Hepatocyte Cells
Classification Serum-free
Form Liquid
Product Type Liver Digest Medium
Serum Level Serum-free
With Additives Phenol Red
Quantity 500 mL
Shipping Condition Dry Ice
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Culture Type Mammalian Cell Culture

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.