Learn More
Thermo Scientific™ Liver Desiccated Bacteriological
Description
Prepare Liver infusion media with Thermo Scientific™ Oxoid™ Liver Desiccated Bacteriological. Liver Desiccated is prepared by the dehydration of fresh pork livers under carefully controlled conditions designed to ensure maximum retention of nutritive properties, and is equivalent to five times its weight of fresh liver
Dehydrated whole liver, specially manufactured for the preparation of infusion media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Appearance | Brown Granules |
| Color | Brown |
| Content And Storage | 10°C to 30°C |
| Description | Liver Desiccated Bacteriological |
| Product Type | Dehydrated Culture Media |
| Quantity | 250 g |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.