missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ LIVE/DEAD™ Cell Vitality Assay Kit, C12 Resazurin/SYTOX™ Green

Product Code. 10174522
Change view
Click to view available options
Quantity:
1000 Assays kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10174522 1000 Assays kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10174522 Supplier Molecular Probes™ Supplier No. L34951

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The LIVE/DEAD Cell Vitality Assay Kit provides a simple, two-color fluorescence assay that distinguishes metabolically active cells from injured cells and dead cells.

  • A simple, two-color fluorescence assay that distinguishes metabolically active cells from injured cells and dead cells
  • Assay is based on the reduction of C12-resazurin to red-fluorescent C12-resorufin in metabolically active cells, and on the uptake of the cell-impermeant, green-fluorescent nucleic acid stain, SYTOX Green dye, in cells with compromised plasma membranes (usually late apoptotic and necrotic cells)
  • In this assay, dead cells emit mostly green fluorescence and healthy, metabolically active cells emit mostly red fluorescence
  • Injured cells exhibit reduced red and green fluorescence
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant, Permeant
Cell Type Mammalian Cells, Eukaryotic Cells
Color Green, Red
Content And Storage Contains 5 vials of C12-resazurin (40 μg each vial), 1 vial of SYTOX™ green (100 μL, 10 μM solution in DMSO), 1 vial of DMSO (1.5 mL), and 10X phosphate buffer (100 mL).

Store in freezer (-5°C to -30°C) and protect from light.
Description LIVE/DEAD™ Cell Vitality Assay Kit, C12 Resazurin/SYTOX™ Green
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer, Microplate Reader
Product Type Cell Vitality Assay Kit
Dye Type Other Label(s) or Dye(s)
Emission C12 Resazurin: 563/587
SYTOX™ Green: 504/523
Excitation Wavelength Range 504/563 nm
Format Tube(s)
Product Line LIVE/DEAD, SYTOX
Quantity 1000 Assays kit
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.