missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIV-1/Zip6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
LIV-1/Zip6 Polyclonal specifically detects LIV-1/Zip6 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | LIV-1/Zip6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | zinc transporter ZIP6, LIV-1, solute carrier family 39 (zinc transporter), member 6 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 170-320 of human LIV-1/Zip6 (NP_001092876.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?