missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIV-1/Zip6 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
£497.00
Specifications
| Antigen | LIV-1/Zip6 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Form | Purified |
Description
LIV-1/Zip6 Polyclonal antibody specifically detects LIV-1/Zip6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| LIV-1/Zip6 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| zinc transporter ZIP6, LIV-1, solute carrier family 39 (zinc transporter), member 6 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of human ZIP6. The immunogen is located within amino acids 430 - 480 of ZIP6. Amino Acid Squence: FKDKKKKNQKKPENDD | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| mFluor Violet 610 SE | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| 50mM Sodium Borate | |
| 25800 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title