missing translation for 'onlineSavingsMsg'
Learn More

Litronesib Racemate, MedChemExpress

Product Code. 16458797
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
This item is not returnable. View return policy

Product Code. 16458797

Brand: Medchem Express HY14846A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Litronesib Racemate (LY2523355 Racemate) is the racemate of litronesib. Litronesib is a selective, allosteric inhibitor of kinesin Eg5.

Litronesib Racemate (LY2523355 Racemate) is the racemate of litronesib. Litronesib is a selective, allosteric inhibitor of kinesin Eg5.

Specifications

Chemical Name or Material Litronesib Racemate
CAS 546111-97-1
Color White
Purity Grade Notes Research
Molecular Formula C23H37N5O4S2
Quantity 5 mg
Synonym LY2523355 Racemate
Solubility Information DMSO : 100 mg/mL (195.43 mM; Need ultrasonic)
SMILES CC(C)(C)C(N1C(C2=CC=CC=C2)(CNS(CCNCC)(=O)=O)SC(NC(C(C)(C)C)=O)=N1)=O
Molecular Weight (g/mol) 511.7
Formula Weight 511.7
Percent Purity 99.2%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.