missing translation for 'onlineSavingsMsg'
Learn More

Litronesib, MedChemExpress

Product Code. 16466566
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16466566 100 mg 100mg
16448797 5 mg 5mg
16442337 50 mg 50mg
16416208 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16466566 Supplier Medchem Express Supplier No. HY14846
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Litronesib (LY2523355) is a selective mitosis-specific kinesin Eg5 inhibitor, with antitumor activity.

Litronesib (LY2523355) is a selective mitosis-specific kinesin Eg5 inhibitor, with antitumor activity.

Specifications

Chemical Name or Material Litronesib
CAS 910634-41-2
Color White
Purity Grade Notes Research
Molecular Formula C23H37N5O4S2
Quantity 100 mg
Synonym LY2523355
Solubility Information DMSO : ≥ 50 mg/mL (97.71 mM)
SMILES CC(C)(C)C(N1[C@@](C2=CC=CC=C2)(CNS(CCNCC)(=O)=O)SC(NC(C(C)(C)C)=O)=N1)=O
Molecular Weight (g/mol) 511.7
Formula Weight 511.7
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.