missing translation for 'onlineSavingsMsg'
Learn More

Lithocholylglycine, TRC

Product Code. 17951265
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17951265 25 mg 25mg
17931695 5 mg 5mg
17944355 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17951265 Supplier Toronto Research Chemicals Supplier No. TRCL46920025MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 474-74-8
Molecular Formula C26 H43 N O4
Molecular Weight (g/mol) 433.62
Synonym Glycine, N-[(3α,5β)-3-hydroxy-24-oxocholan-24-yl]-, Glycine, N-(3α-hydroxy-5β-cholan-24-oyl)- (8CI), Glycolithocholic acid (6CI,7CI), 5β-Cholan-24-amide, N-(carboxymethyl)-3α-hydroxy- (8CI), Cholane, glycine deriv., N-[(3α,5β)-3-Hydroxy-24-oxocholan-24-yl]glycine, 3α-Hydroxy-5β-cholanic acid glycine ester, 3α-Hydroxy-5β-cholanoylglycine, 3α-Hydroxy-N-(carboxymethyl)-5β-cholan-24-amide, Lithocholic acid glycine conjugate, Lithocholylglycine, N-(3α-Hydroxy-5β-cholanoyl)glycine
IUPAC Name 2-[[(4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid
SMILES C[C@H](CCC(=O)NCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C26H43NO4/c1-16(4-9-23(29)27-15-24(30)31)20-7-8-21-19-6-5-17-14-18(28)10-12-25(17,2)22(19)11-13-26(20,21)3/h16-22,28H,4-15H2,1-3H3,(H,27,29)(H,30,31)/t16-,17-,18-,19+,20-,21+,22+,25+,26-/m1/s1
Formula Weight 433.3192
Percent Purity >95%
Chemical Name or Material Lithocholylglycine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.