missing translation for 'onlineSavingsMsg'
Learn More

Lithium zirconium oxide, 94% min, Thermo Scientific Chemicals

Product Code. 11348148
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348148 25 g 25g
11358148 100 g 100g
11368148 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11348148 Supplier Thermo Scientific Alfa Aesar Supplier No. 039377.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lithium zirconium oxide is used in the separation and absorption of carbon dioxide. It is used in the manufacture of dielectric bodies and insulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12031-83-3
Molecular Formula Li2O3Zr
Molecular Weight (g/mol) 153.10
MDL Number MFCD00016182
InChI Key PSKLXRHTVCSWPJ-UHFFFAOYSA-N
Synonym lithium zirconate, dilithium dioxido oxo zirconium, 2li.o3zr, lithium zirconium oxide, dilithotab oxozirconiumbis olate, lithium zirconium oxide min, dilithium 1+ ion oxozirconiumbis olate, dilithium 1+ oxozirconiumbis olate
PubChem CID 6093659
IUPAC Name dilithium;dioxido(oxo)zirconium
SMILES [Li+].[Li+].[O-][Zr]([O-])=O

Specifications

Color White to Cream
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 153.1
Percent Purity ≤94%
Odor Odorless
Chemical Name or Material Lithium zirconium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.