missing translation for 'onlineSavingsMsg'
Learn More

Lithium tungsten oxide, 99% min

Product Code. 11339548
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339548 50 g 50g
11349548 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339548 Supplier Thermo Scientific Alfa Aesar Supplier No. 043366.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13568-45-1 | Li2O4W | 261.716 g/mol

Lithium tungsten oxide is employed for the preparation of ceramics with ultra-low sintering temperatures. It acts as a catalyst for oxidative coupling reactions. Furthermore, it is used in the production of high density aqueous polytungstate (metatungstate) solutions which are utilized for the separation of minerals and other solids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13568-45-1
Molecular Formula Li2O4W
Molecular Weight (g/mol) 261.716
MDL Number MFCD00016185
InChI Key SCKKBXOJPMZVPF-UHFFFAOYSA-N
Synonym lithium tungstate, dilithium dioxido dioxo tungsten, dilithotab tungstate, 2li.wo4, dilithium 1+ ion tungstate
PubChem CID 6095624
IUPAC Name dilithium;dioxido(dioxo)tungsten
SMILES [Li+].[Li+].[O-][W](=O)(=O)[O-]

Specifications

Melting Point 742°C
Physical Form Powder
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 261.73
Percent Purity ≥99%
Density 3.71 g/mL
Chemical Name or Material Lithium tungsten oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.