Learn More
Lithium tungsten oxide, 99% min
Description
Lithium tungsten oxide is employed for the preparation of ceramics with ultra-low sintering temperatures. It acts as a catalyst for oxidative coupling reactions. Furthermore, it is used in the production of high density aqueous polytungstate (metatungstate) solutions which are utilized for the separation of minerals and other solids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 742°C |
| Physical Form | Powder |
| Quantity | 250 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 261.73 |
| Percent Purity | ≥99% |
| Density | 3.71 g/mL |
| Chemical Name or Material | Lithium tungsten oxide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.