missing translation for 'onlineSavingsMsg'
Learn More

Lithium (trimethylsilyl)acetylide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10310524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
AcroSeal™ Glass Bottle in Tin Can
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10310524 800 mL AcroSeal™ Glass Bottle 800mL
10359213 100 mL AcroSeal™ Glass Bottle in Tin Can 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10310524 Supplier Thermo Scientific Acros Supplier No. 380658000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54655-07-1 , 109-99-9
Molecular Formula C5H9LiSi
Molecular Weight (g/mol) 104.15
MDL Number MFCD00075059
InChI Key WDDOQHLJFOUQMW-UHFFFAOYSA-N
Synonym trimethylsilyl ethynyllithium, lithium, trimethylsilyl ethynyl, lithium 1+ ion trimethylsilyl ethyne, lithium trimethylsilylacetylide, lithio trimethylsilyl acetylene, lithium trimethylsilyl acetylide, lithium ethynyl trimethyl silane, lithium trimethylsilylacetylenide, zvxxeonxfwsciz-uhfffaoysa-n
PubChem CID 3431600
IUPAC Name lithium;ethynyl(trimethyl)silane
SMILES [Li+].C[Si](C)(C)C#[C-]

Specifications

Color Yellow to Orange
Physical Form Solution
Quantity 800 mL
Solubility Information Solubility in water: reacts
Formula Weight 104.15
Concentration or Composition (by Analyte or Components) 0.4 to 0.6M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.88
Density 0.8800g/mL
Chemical Name or Material Lithium (trimethylsilyl)acetylide
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium (trimethylsilyl)acetylide, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.