missing translation for 'onlineSavingsMsg'
Learn More

Lithium trifluoromethanesulfonate, 98%

Product Code. 10445282 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10445282 25 g Glass bottle 25g
10068771 5 g Glass bottle 5g
10521384 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10445282 Supplier Thermo Scientific Acros Supplier No. 358710250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33454-82-9 | CF3LiO3S | 156.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33454-82-9
Molecular Formula CF3LiO3S
Molecular Weight (g/mol) 156.01
MDL Number MFCD00013227
InChI Key MCVFFRWZNYZUIJ-UHFFFAOYSA-M
Synonym lithium trifluoromethanesulfonate, lithium triflate, li triflate, lithium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, lithium salt, unii-1c966kv50i, trifluoromethanesulfonic acid lithium salt, lithium 1+ ion trifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, lithium salt 1:1, trifluoromethanesulfonic acid, lithium salt
PubChem CID 3664839
IUPAC Name lithium;trifluoromethanesulfonate
SMILES [Li+].C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 423.0°C
Color White or Colorless
Infrared Spectrum Authentic
Assay Percent Range 4.34 to 4.56% (Li)
Packaging Glass bottle
Linear Formula F3CSO3Li
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 156.01
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Lithium trifluoromethanesulfonate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Lithium trifluoromethanesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.