missing translation for 'onlineSavingsMsg'
Learn More

Lithium trifluoromethanesulfonate, 97%

Product Code. 11323738
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323738 10 g 10g
11333738 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323738 Supplier Thermo Scientific Alfa Aesar Supplier No. 039322.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33454-82-9 | CF3LiO3S | 156.003 g/mol

Lithium trifluoromethanesulfonate is mixed with ceramic filler to prepare polyethylene oxide films in order to improve the electrochemical and mechanical characteristics of the membrane, which is used in the lithium batteries. It acts as a doping salt used in the preparation of nano manganese-composite polymer electrolytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33454-82-9
Molecular Formula CF3LiO3S
Molecular Weight (g/mol) 156.003
MDL Number MFCD00013227
InChI Key MCVFFRWZNYZUIJ-UHFFFAOYSA-M
Synonym lithium trifluoromethanesulfonate, lithium triflate, li triflate, lithium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, lithium salt, unii-1c966kv50i, trifluoromethanesulfonic acid lithium salt, lithium 1+ ion trifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, lithium salt 1:1, trifluoromethanesulfonic acid, lithium salt
PubChem CID 3664839
IUPAC Name lithium;trifluoromethanesulfonate
SMILES [Li+].C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 423°C
Color White
Quantity 10 g
Sensitivity Moisture sensitive
Solubility Information Miscible with water.
Formula Weight 156.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Lithium trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.