missing translation for 'onlineSavingsMsg'
Learn More

Lithium Tetrafluoroborate, MP Biomedicals

Product Code. 11448465 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448465 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11448465 Supplier MP Biomedicals Supplier No. 0522108810
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Chemical Name: Lithium tetrafluoroborate
  • Molecular Formula: BF4Li
  • Formula Weight: 93.74
  • CAS Number: 14283-07-9

Specifications

CAS 14283-07-9
Quantity 10 g
Sensitivity Hygroscopic
Molecular Formula BF4Li
Linear Formula LiBF4
MDL Number MFCD00011087
Synonym lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n
InChI Formula InChI=1S/BF4.Li/c2-1(3,4)5;/q-1;+1
InChI Key UFXJWFBILHTTET-UHFFFAOYSA-N
SMILES [Li+].F[B-](F)(F)F
IUPAC Name lithium(1+) tetrafluoroboranuide
Molecular Weight (g/mol) 93.74
PubChem CID 4298216
Formula Weight 93.75
Chemical Name or Material Lithium Tetra Fluoroborate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.