missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetrafluoroborate, 98%

Product Code. 11317217
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317217 50 g 50g
11307217 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317217 Supplier Thermo Scientific Alfa Aesar Supplier No. 011528.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14283-07-9 | BF4Li | 93.74 g/mol

Lithium tetrafluoroborate is extensively utilized as an electrolyte in primary and secondary cells. It is used in rechargeable Li ion batteries. The electrolytes are non aqueous solutions of lithium tetrafluoroborate in the organic media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14283-07-9
Molecular Formula BF4Li
Molecular Weight (g/mol) 93.74
MDL Number MFCD00011087
InChI Key UFXJWFBILHTTET-UHFFFAOYSA-N
Synonym lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n
PubChem CID 4298216
IUPAC Name lithium;tetrafluoroborate
SMILES [Li+].F[B-](F)(F)F

Specifications

Melting Point 293°C to 300°C (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Odor Odorless
Sensitivity Very Hygroscopic
Linear Formula LiBF4
UN Number UN3260
Merck Index 14,5543
Solubility Information Soluble in water and tetrahydrofuran. Slightly soluble is diethyl ether.
Formula Weight 93.75
Percent Purity 98%
Chemical Name or Material Lithium tetrafluoroborate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.