missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetrafluoroborate, 98%, pure

Product Code. 10358550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10358550 50 g Plastic bottle 50g
10032880 10 g Plastic bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10358550 Supplier Thermo Scientific Acros Supplier No. 223770500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14283-07-9 | BF4Li | 93.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14283-07-9
Molecular Formula BF4Li
Molecular Weight (g/mol) 93.74
MDL Number MFCD00011087
InChI Key UFXJWFBILHTTET-UHFFFAOYSA-N
Synonym lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n
PubChem CID 4298216
IUPAC Name lithium(1+) tetrafluoroboranuide
SMILES [Li+].F[B-](F)(F)F

Specifications

Melting Point 275°C
Color Gray to White
Physical Form Crystalline Powder and/or Crystals
Quantity 50 g
Packaging Plastic bottle
Specific Gravity 0.852
Assay Percent Range 98%
Density 0.8520g/mL
Linear Formula LiBF4
Fieser 05,416; 10,248
Merck Index 15, 5599
Solubility Information Solubility in water: soluble
Formula Weight 93.75
Percent Purity 98%
Grade Pure
Chemical Name or Material Lithium tetrafluoroborate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium tetrafluoroborate, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.