missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Product Code. 11307217
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Lithium tetrafluoroborate is extensively utilized as an electrolyte in primary and secondary cells. It is used in rechargeable Li ion batteries. The electrolytes are non aqueous solutions of lithium tetrafluoroborate in the organic media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14283-07-9
Molecular Formula BF4Li
Molecular Weight (g/mol) 93.74
MDL Number MFCD00011087
InChI Key UFXJWFBILHTTET-UHFFFAOYSA-N
Synonym lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n
PubChem CID 4298216
IUPAC Name lithium;tetrafluoroborate
SMILES [Li+].F[B-](F)(F)F

Specifications

Melting Point 293°C to 300°C (decomposition)
Color White
Physical Form Powder
Quantity 10 g
Linear Formula LiBF4
UN Number UN3260
Merck Index 14,5543
Solubility Information Soluble in water and tetrahydrofuran. Slightly soluble is diethyl ether.
Formula Weight 93.75
Percent Purity 98%
Odor Odorless
Sensitivity Very Hygroscopic
Chemical Name or Material Lithium tetrafluoroborate
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'