missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetrachlorocuprate, 0.1M in THF

Product Code. 11480500
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480500 500 mL 500mL
11367957 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480500 Supplier Thermo Scientific Alfa Aesar Supplier No. 020620.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15489-27-7 | Cl4CuLi2 | 219.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15489-27-7
Molecular Formula Cl4CuLi2
Molecular Weight (g/mol) 219.226
MDL Number MFCD00011081
InChI Key HCJWWBBBSCXJMS-UHFFFAOYSA-J
PubChem CID 11074879
IUPAC Name dilithium;tetrachlorocopper(2-)
SMILES [Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl

Specifications

Color Orange
Physical Form Liquid
Quantity 500 mL
UN Number UN2056
Solubility Information Difficult to mix.
Formula Weight 219.24
Concentration or Composition (by Analyte or Components) 0.1M in THF
Sensitivity Moisture sensitive
Density 0.910 g/mL
Chemical Name or Material Lithium tetrachlorocuprate

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.