missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetraborate trihydrate, 96%, Thermo Scientific Chemicals

£26.40 - £98.50

Chemical Identifiers

CAS 37195-62-3
Molecular Formula Li2B4O7·3H2O
MDL Number MFCD00167253
Synonym lithium borate trihydrate, acmc-1ajhy, trilithium 1+ trihydrate borate, trilithium 1+ ion trihydrate borate
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11494153
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10358.22
100 g
£26.40
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11404163
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10358.36
500 g
£98.50
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is employed in the conductances of some potassium and sodium salts in dimethylformamide at 25°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the conductances of some potassium and sodium salts in dimethylformamide at 25°C.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

37195-62-3
Li2B4O7·3H2O
MFCD00167253
lithium borate trihydrate, acmc-1ajhy, trilithium 1+ trihydrate borate, trilithium 1+ ion trihydrate borate

Specifications

37195-62-3
MFCD00167253
lithium borate trihydrate, acmc-1ajhy, trilithium 1+ trihydrate borate, trilithium 1+ ion trihydrate borate
223.17 (169.12 Anhydrous)
Lithium tetraborate trihydrate
Li2B4O7·3H2O
14,5524
Soluble in water.
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 234-514-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P235-Keep cool.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only