missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetra(2-methyl-8-hydroxyquinolinato)boron, 98%, Thermo Scientific™

Product Code. 15442116
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338949-42-1
Molecular Formula C40H32BLiN4O4
Molecular Weight (g/mol) 650.47
MDL Number MFCD03093696
InChI Key AVPRLXSPVRRDIE-UHFFFAOYSA-N
Synonym lithium tetra 2-methyl-8-hydroxyquinolinato boron, lithium tetra 2-methyl-8, lithium tetrakis 2-methylquinolin-8-olato-kappao borate 1-, lithium 1+ tetrakis 2-methylquinolin-8-yl oxy boranuide
PubChem CID 23714786
IUPAC Name lithium;tetrakis[(2-methylquinolin-8-yl)oxy]boranuide
SMILES [Li+].[B-](OC1=CC=CC2=C1N=C(C=C2)C)(OC3=CC=CC4=C3N=C(C=C4)C)(OC5=CC=CC6=C5N=C(C=C6)C)OC7=CC=CC8=C7N=C(C=C8)C

Specifications

Melting Point 250°C to 258°C
Color White
Physical Form Solid
Quantity 100 mg
Formula Weight 650.46
Percent Purity 98%
Chemical Name or Material Lithium tetra(2-methyl-8-hydroxyquinolinato)boron
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'