missing translation for 'onlineSavingsMsg'
Learn More

Lithium tert-butoxide, 99.9% (metals basis)

Product Code. 11369618
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Unit Size:
10g
250g
2g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369618 50 g 50g
11389618 2 g 2g
11379618 10 g 10g
11399618 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11369618 Supplier Thermo Scientific Alfa Aesar Supplier No. 044133.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1907-33-1 | C4H9LiO | 80.055 g/mol

Lithium tert-butoxide is a mild base used in organic synthesis. It is involved in the alfa-alkylation reaction of ketones with primary alcohols. It is utilized as a catalyst in the reduction of ketones to the corresponding secondary alcohols. It acts as lithium source utilized in the preparation of LixTiyOz thin films. Further, it is used in conjunction with organometallic reagents/catalysts to modify their reactivity and selectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1907-33-1
Molecular Formula C4H9LiO
Molecular Weight (g/mol) 80.055
MDL Number MFCD00050479
InChI Key LZWQNOHZMQIFBX-UHFFFAOYSA-N
Synonym lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate
PubChem CID 23664764
IUPAC Name lithium;2-methylpropan-2-olate
SMILES [Li+].CC(C)(C)[O-]

Specifications

Melting Point ∼150°C (decomposition)
Color White to Cream
Odor Odorless
Linear Formula (CH3)3COLi
Quantity 50 g
UN Number UN3206
Beilstein 3620018
Sensitivity Air and moisture sensitive
Solubility Information Soluble in toluene,hexane,tetrahydrofuran and methyl tert-butyl ether.
Formula Weight 80.06
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Lithium tert-butoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.