Learn More
Lithium tert-butoxide, 99.9% (metals basis)
Description
Lithium tert-butoxide is a mild base used in organic synthesis. It is involved in the alfa-alkylation reaction of ketones with primary alcohols. It is utilized as a catalyst in the reduction of ketones to the corresponding secondary alcohols. It acts as lithium source utilized in the preparation of LixTiyOz thin films. Further, it is used in conjunction with organometallic reagents/catalysts to modify their reactivity and selectivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼150°C (decomposition) |
| Color | White to Cream |
| Odor | Odorless |
| Linear Formula | (CH3)3COLi |
| Quantity | 10 g |
| UN Number | UN3206 |
| Beilstein | 3620018 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Soluble in toluene,hexane,tetrahydrofuran and methyl tert-butyl ether. |
| Formula Weight | 80.06 |
| Show More |
Safety and Handling
- Lithium tert-butoxide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH014-Reacts violently with water.
- P235+P410-Keep cool. Protect from sunlight.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Lithium tert-butoxide
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.