missing translation for 'onlineSavingsMsg'
Learn More

Lithium tantalum ethoxide, 99.9% (metals basis), 5% w/v in ethanol, Thermo Scientific™

Catalog Number 11374438
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
127503-04-2
MFCD00210630
lithium tantalum ethoxide
This item is not returnable. View return policy
127503-04-2
MFCD00210630
lithium tantalum ethoxide

Precursor for Lithium tantalum oxide films

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Yellow
Odor Alcohol-like
Linear Formula LiTa(OCH2CH3)6
Quantity 100 mL
UN Number UN1170
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 458.04
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Lithium tantalum ethoxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'