Learn More
Lithium sulfate monohydrate, ACS, 99.0% min
Description
Lithium sulfate monohydrate is used in solar panels and potential for a new class of battery. It is also used to treat bipolar disorder and in ultrasound-type non-destructive testing. It is also used as an additive for portland cement to speed up the curing. It is also involved as a catalyst in the elimination reaction for the conversion of n-butyl bromide to 1-butene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 130°C-H2O |
| Color | Colorless |
| Physical Form | Crystalline |
| Quantity | 500 g |
| Assay Percent Range | 99.0% min. |
| Solubility Information | Soluble in water. Soluble in water. Insoluble in ethanol,acetone and pyridine. |
| IUPAC Name | dilithium(1+) hydrate sulfate |
| Formula Weight | 127.96 (109.94 Anhydrous) |
| Grade | ACS |
| Odor | Odorless |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.