missing translation for 'onlineSavingsMsg'
Learn More

Lithium sulfate, anhydrous, 99.7% (metals basis)

Product Code. 11396718
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 11396718

Brand: Thermo Scientific Alfa Aesar 013404.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10377-48-7 | Li2O4S | 109.94 g/mol

Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-48-7
Molecular Formula Li2O4S
Molecular Weight (g/mol) 109.94
MDL Number MFCD00011086
InChI Key INHCSSUBVCNVSK-UHFFFAOYSA-L
Synonym lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
PubChem CID 66320
ChEBI CHEBI:53474
SMILES [Li+].[Li+].[O-]S([O-])(=O)=O

Specifications

Melting Point 845°C
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.7% (Metal basis)
Solubility Information Soluble in water. Insoluble in acetone,ethanol and pyridine.
IUPAC Name dilithium(1+) sulfate
Formula Weight 109.94
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 2.22
Chemical Name or Material Lithium sulfate, anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.