missing translation for 'onlineSavingsMsg'
Learn More

Lithium sulfate, anhydrous, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog Number 11386718
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
10377-48-7
Li2O4S
109.94
MFCD00011086
INHCSSUBVCNVSK-UHFFFAOYSA-L
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
66320
CHEBI:53474
[Li+].[Li+].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
10377-48-7
Li2O4S
109.94
MFCD00011086
INHCSSUBVCNVSK-UHFFFAOYSA-L
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
66320
CHEBI:53474
[Li+].[Li+].[O-]S([O-])(=O)=O

Lithium sulfate is used in the treatment of bipolar disorder. It acts as an important component of ion conducting glasses, solar panels and in battery. It serves as an additive for Portland cement. Further, it is used in the production of special high-strength glass, food and flavoring agent in synthetic beverages. In addition to this, it is used as a catalyst in organic synthesis for the preparation of 1-butene from n-butyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 845°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.7% (Metal basis)
Solubility Information Soluble in water. Insoluble in acetone,ethanol and pyridine.
IUPAC Name dilithium(1+) sulfate
Formula Weight 109.94
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 2.22 g/cm3 at 20°C
Chemical Name or Material Lithium sulfate, anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'