missing translation for 'onlineSavingsMsg'
Learn More

Lithium sulfate, 99%, extra pure, anhydrous

Product Code. 15139243 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 15139243

Brand: Thermo Scientific Acros 218320051

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10377-48-7 | Li2O4S | 109.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-48-7
Molecular Formula Li2O4S
Molecular Weight (g/mol) 109.94
MDL Number MFCD00011086
InChI Key INHCSSUBVCNVSK-UHFFFAOYSA-L
Synonym lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
PubChem CID 66320
ChEBI CHEBI:53474
SMILES [Li+].[Li+].[O-]S([O-])(=O)=O

Specifications

Melting Point 859.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 kg
Assay Percent Range 99%
Linear Formula Li2SO4
Merck Index 15,5597
Solubility Information Solubility in water: 25.7 g/100mL (25°C). Other solubilities: practically insoluble in alcohol
IUPAC Name dilithium;sulfate
Formula Weight 109.94
Percent Purity 99%
Grade Extra Pure
Packaging Plastic Drum
Specific Gravity 2.22
Density 2.2200g/mL
Chemical Name or Material Lithium sulfate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Lithium sulfate, anhydrous
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.