missing translation for 'onlineSavingsMsg'
Learn More

Lithium stearate

Product Code. 11388098
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388098 1 kg 1kg
11398098 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388098 Supplier Thermo Scientific Alfa Aesar Supplier No. 039335.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4485-12-5 | C9H11ClN3 | 196.66 g/mol

Lithium stearate is used as a thickener and gelling agent to prepare oils into lubricating greases. It is the active component of lithium grease. Further, it is used as lubricants, lubricant additives, plastic and rubber products. In addition to this, it is used as a stabilizing agent in the plastics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4485-12-5
MDL Number MFCD00042032

Specifications

Melting Point 220°C
Molecular Formula C18H35LiO2
Quantity 1 kg
Synonym lithium stearate, lithium octadecanoate, octadecanoic acid, lithium salt, stearic acid, lithium salt, lithalure, litholite, stavinor, lithium stearate, pure, stearic acid lithium salt, unii-p31mc94p70
Solubility Information Insoluble in water.
InChI Key HGPXWXLYXNVULB-UHFFFAOYSA-M
SMILES [Li+].CCCCCCCCCCCCCCCCCC(=O)[O-]
IUPAC Name lithium;octadecanoate
Molecular Weight (g/mol) 290.42
PubChem CID 517357
Formula Weight 290.41
Physical Form Powder
Chemical Name or Material Lithium stearate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.