missing translation for 'onlineSavingsMsg'
Learn More

Lithium Standard: Li @ 1000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 17980876
Change view
Click to view available options
Quantity:
50 g
200 g
Unit Size:
200g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17980876 200 g 200g
16419659 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17980876 Supplier VHG Labs Supplier No. VHGOLI1000200G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Lithium Standard: Li @ 1000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Lithium
CAS 7439-93-2
Methods ICP/XRF/RDE
Analyte or Component Names Lithium
Shipping Condition Room Temperature
Molecular Formula Li
Certifications/Compliance ISO 17034
Synonym Li
InChI Formula InChI=1S/Li
SMILES [Li]
IUPAC Name Lithium
Solvent or Matrix 75 cSt Mineral Oil
Molecular Weight (g/mol) 6.94
Quantity 200 g
Formula Weight 7.016 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.