missing translation for 'onlineSavingsMsg'
Learn More

Lithium Standard For ICP, 1000μg/mL in 2% HCl, SPEX CertiPrep™

Codice prodotto. 10316082 Sfoglia Tutto SPEX Certiprep Prodotti
Cambia vista
Click to view available options
Quantity:
500 mL
Dimensione della confezione:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
10316082 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10316082 Fornitore SPEX Certiprep N. del fornitore PLLI12X
  • ONLINE EXCLUSIVE PRICE
    £160.82
    £132.82 / 500mL
    Save £28.00
    17% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Li1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifica

Packaging Triple-leached LDPE bottle
CAS 7439-93-2, 7647-01-0, 7732-18-5
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
CAS Min % 1.0, 5.0, 94.0
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 1.0, 5.0, 94.0
Molecular Formula Li
MDL Number MFCD00134051
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key WHXSMMKQMYFTQS-UHFFFAOYSA-N
Type Certified Reference Material
SMILES [Li]
IUPAC Name lithium
Solvent or Matrix 2% HCl
Molecular Weight (g/mol) 6.94
PubChem CID 3028194
Quantity 500 mL
ChEBI CHEBI:30145
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLLI2-2Y, -2X    10,000    PLLI2-3Y, -3X
2-5% HCl    1000μg/mL    PLLI1-2X    10,000    PLLI1-3X
Chemical Name or Material Lithium Standard
Vedi altri risultati Mostra meno risultati

Product intended for laboratory use only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.