missing translation for 'onlineSavingsMsg'
Learn More

Lithium Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™

Product Code. 16321206 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16321206 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16321206 Supplier SPEX Certiprep Supplier No. ORGLI82Z
  • ONLINE EXCLUSIVE PRICE
    £75.62
    £61.72 / 50g
    Save £13.90
    18% Off
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Li1000μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 7439-93-2, 8042-47-5
Certificate of Analysis With Certificate of Analysis.
CAS Min % 0.1, 99.9
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.1, 99.9
Molecular Formula Li
MDL Number MFCD00134051
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key WHXSMMKQMYFTQS-UHFFFAOYSA-N
Type Certified Reference Material
SMILES [Li]
IUPAC Name lithium
Solvent or Matrix Base Oil 20
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 6.94
PubChem CID 3028194
Quantity 50 g
ChEBI CHEBI:30145
Grade Oil/Plasma
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
Base Oil 75 1000μg/g ORG-LI8-2Z
Chemical Name or Material Lithium Oil Standard
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.