missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lithium pyruvate monohydrate

Product Code. 11437777
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437777 50 g 50g
11427777 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11437777 Supplier Alfa Aesar™ Supplier No. A16308.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Identificadores de productos químicos

CAS 2922-61-4
Molecular Formula C3H3LiO3
Molecular Weight (g/mol) 93.994
MDL Number MFCD00150352
InChI Key OFJHGWPRBMPXCX-UHFFFAOYSA-M
Synonym lithium pyruvate, lithium 2-oxopropanoate, pyruvic acid lithium salt, propanoic acid, 2-oxo-, lithium salt, lithiumpyruvat, lithotab 2-oxopropanoate, propanoic acid, 2-oxo-, lithium salt 1:1, c3h3o3.li, lithium pyruvate h2o, lithium 1+ pyruvate
PubChem CID 23663618
IUPAC Name lithium;2-oxopropanoate
SMILES [Li+].CC(=O)C(=O)[O-]

Especificaciones

Quantity 50 g
Formula Weight 112.01 (94.00 Anhydrous)
Chemical Name or Material Lithium pyruvate monohydrate
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.