missing translation for 'onlineSavingsMsg'
Learn More

Lithium phosphate, Puratronic™, 99.99% (metals basis)

Product Code. 11396827
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396827 100 g 100g
11386827 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396827 Supplier Thermo Scientific Alfa Aesar Supplier No. 010743.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10377-52-3 | Li3O4P | 115.79 g/mol

Lithium phosphate batteries batteries are now widely used as a staple for cell phones, laptops and some other gadgets. Used in low-expansion porcelain enamel glazes and polymer intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-52-3
Molecular Formula Li3O4P
Molecular Weight (g/mol) 115.79
MDL Number MFCD00016187
InChI Key TWQULNDIKKJZPH-UHFFFAOYSA-K
Synonym lithium phosphate, lithium orthophosphate, trilithium orthophosphate, trilithium phosphate, phosphoric acid, trilithium salt, unii-2qm4k05q74, lithiumphosphate, trilithotab phosphate, phosphoric acid, lithium salt 1:3, li3po4
PubChem CID 165867
SMILES [Li+].[Li+].[Li+].[O-]P([O-])([O-])=O

Specifications

Melting Point 837°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Poor solubility in water. Insoluble in alcohols.
IUPAC Name trilithium;phosphate
Formula Weight 115.79
Percent Purity 99.99%
Grade Puratronic
Odor Odorless
Density 2.54
Assay (metals basis)
Chemical Name or Material Lithium phosphate
Show More Show Less
compliance-icons
Product Identifier
  • Lithium phosphate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Lithium phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.