missing translation for 'onlineSavingsMsg'
Learn More

Lithium phosphate, extra pure, Thermo Scientific Chemicals

Product Code. 10569413
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-52-3
Molecular Formula Li3O4P
Molecular Weight (g/mol) 115.79
MDL Number MFCD00016187
InChI Key TWQULNDIKKJZPH-UHFFFAOYSA-K
Synonym lithium phosphate, lithium orthophosphate, trilithium orthophosphate, trilithium phosphate, phosphoric acid, trilithium salt, unii-2qm4k05q74, lithiumphosphate, trilithotab phosphate, phosphoric acid, lithium salt 1:3, li3po4
PubChem CID 165867
SMILES [Li+].[Li+].[Li+].[O-]P([O-])([O-])=O

Specifications

Melting Point 837°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Linear Formula Li3PO4
Solubility Information Solubility in water: 0.4g/L (20°C)
IUPAC Name trilithium(1+) phosphate
Formula Weight 115.79
Percent Purity ≥95.0%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Lithium phosphate
Show More Show Less
compliance-icons
Product Identifier
  • Lithium phosphate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'