missing translation for 'onlineSavingsMsg'
Learn More

Lithium peroxide, 95%, Thermo Scientific™

Product Code. 17241152 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Plastic Bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
17241152 10 g Plastic Bottle 10g
10761135 50 g Plastic Bottle 50g
10793245 250 g Plastic Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17241152 Supplier Thermo Scientific Acros Supplier No. 389880100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12031-80-0
Molecular Formula Li2O2
Molecular Weight (g/mol) 45.88
MDL Number MFCD00016184
InChI Key HPGPEWYJWRWDTP-UHFFFAOYSA-N
Synonym lithium peroxide li2 o2, lithium peroxide un1472 oxidizer, lithium oxide, dilithium peroxide, unii-9anx556r5f
PubChem CID 25489
IUPAC Name dilithium;peroxide
SMILES [Li+].[Li+].[O-][O-]

Specifications

Color White to Yellow
Physical Form Powder
Quantity 10 g
Assay Percent Range 95%
Linear Formula LiO2Li
Formula Weight 45.88
Percent Purity 95%
Packaging Plastic Bottle
Chemical Name or Material Lithium peroxide
compliance-icons compliance-icons
Product Identifier
  • Lithium peroxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Lithium peroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.