missing translation for 'onlineSavingsMsg'
Learn More

Lithium peroxide, 95%

Product Code. 11326758
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326758 500 g 500g
11316758 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326758 Supplier Thermo Scientific Alfa Aesar Supplier No. 013421.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12031-80-0 | Li2O2 | 45.878 g/mol

Lithium peroxide is used in air purifiers. It is also used to produce high-purity oxygen. It acts as a catalyst for polymerization of styrene to polystyrene. Further, it serves as a curing agent for special polymers and a source of oxygen in sealed spaces such as submarines and in breathing apparatus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12031-80-0
Molecular Formula Li2O2
MDL Number MFCD00016184
InChI Key HPGPEWYJWRWDTP-UHFFFAOYSA-N
PubChem CID 25489
IUPAC Name dilithium;peroxide
SMILES [Li+].[Li+].[O-][O-]

Specifications

Color White
Physical Form Powder
Quantity 500 g
UN Number UN1472
Synonym dilithium peroxide, lithium peroxide li2 o2, lithium oxide, unii-9anx556r5f, lithium peroxide un1472 oxidizer
Solubility Information Soluble in anhydrous acetic acid. Insoluble in alcohol.
Molecular Weight (g/mol) 45.878
Formula Weight 45.88
Percent Purity 95%
Odor Odorless
Sensitivity Air and moisture sensitive
Chemical Name or Material Lithium peroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.