missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium peroxide, 95%, Thermo Scientific Chemicals
204.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Description
Lithium peroxide is used in air purifiers. It is also used to produce high-purity oxygen. It acts as a catalyst for polymerization of styrene to polystyrene. Further, it serves as a curing agent for special polymers and a source of oxygen in sealed spaces such as submarines and in breathing apparatus.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 100 g |
UN Number | UN1472 |
Synonym | dilithium peroxide, lithium peroxide li2 o2, lithium oxide, unii-9anx556r5f, lithium peroxide un1472 oxidizer |
Solubility Information | Soluble in anhydrous acetic acid. Insoluble in alcohol. |
Molecular Weight (g/mol) | 45.878 |
Formula Weight | 45.88 |
Percent Purity | 95% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'