missing translation for 'onlineSavingsMsg'
Learn More

Lithium periodate dihydrate, Reagent Grade

Product Code. 11338118
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338118 250 g 250g
11328118 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338118 Supplier Thermo Scientific Alfa Aesar Supplier No. 039355.30
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 307310-74-3 | ILiO4 | 197.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307310-74-3
Molecular Formula ILiO4
Molecular Weight (g/mol) 197.84
MDL Number MFCD00049474
InChI Key SYWXNZXEJFSLEU-UHFFFAOYSA-M
Synonym lithium periodate, lithotab io4, acmc-20akme, lithium 1+ ion periodate, lithium 1+ periodate, lithium periodate lih4io6
PubChem CID 23711816
IUPAC Name lithium;periodate
SMILES [Li+].[O-]I(=O)(=O)=O

Specifications

Melting Point >370°C
Physical Form Powder
Quantity 250 g
Odor Odorless
UN Number UN1479
Solubility Information Soluble in water.
Formula Weight 233.87 (197.84 Anhydrous)
Grade Reagent
Chemical Name or Material Lithium periodate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.