missing translation for 'onlineSavingsMsg'
Learn More

Lithium Perchlorate, ACS Reagent, ≥95.0%, Honeywell™ Fluka™

Product Code. 15611760 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15611760 500 g Poly Bottle 500g
15621760 5 g Poly Bottle 5g
15601760 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15611760 Supplier Honeywell-Fluka Supplier No. 205281500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent,≥95.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3-9-7791
Molecular Formula ClH6LiO7
Molecular Weight (g/mol) 160.43
MDL Number MFCD00011079
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M

Specifications

Melting Point 236°C
Color White
pH 6.0 to 9.5 (at 106.4g/L 25°C)
Quantity 500 g
Linear Formula LiClO4
UN Number UN1481
Synonym sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
SMILES [Li+].O.O.O.[O-][Cl](=O)(=O)=O
IUPAC Name sodium;perchlorate;hydrate
PubChem CID 516899
Formula Weight 106.39g/mol
Percent Purity ≥95.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Granules
Density 2.420g/cu.cm
Chemical Name or Material Lithium Perchlorate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.