missing translation for 'onlineSavingsMsg'
Learn More

Lithium oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number 11384787
Click to view available options
Quantity:
10 g
50 g
250 g
12057-24-8
Li2O
MFCD00016183
12057-24-8
Li2O
MFCD00016183

For Use As a Ceramics and Special Glass Formulations, Carbon Dioxide Absorbent

Ceramics and special glass formulations, carbon dioxide absorbent.Lithium oxide is used as a flux in ceramic glazes, co-dopant with yttria in the zirconia ceramic top coat and as the cathode in the lithium ion batteries, which is used in power electronic devices like mobile phones, laptop computers and battery-powered cars. It is also used to prepare lithium hydroxide and lithium metal by electrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Air and Moisture Sensitive; Readily absorbs carbon dioxide and water from air
TRUSTED_SUSTAINABILITY
Melting Point 1427°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.5% (Metal basis)
Solubility Information Soluble in water.
Formula Weight 29.88
Odor Odorless
Sensitivity Air and moisture sensitive
Appearance White powder
Density 2.013 g/cm3 at 20°C
Chemical Name or Material Lithium oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'