missing translation for 'onlineSavingsMsg'
Learn More

Lithium oxide, 99.5% (metals basis)

Product Code. 11394787
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11394787

Brand: Thermo Scientific Alfa Aesar 041832.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As a Ceramics and Special Glass Formulations, Carbon Dioxide Absorbent | CAS: 12057-24-8 | Li2O

The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher, and to treat hypokalemia with its pharmaceutical preparation, a condition in which the level of potassium will be low in blood. Further it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Air and Moisture Sensitive; Readily absorbs carbon dioxide and water from air
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12057-24-8
Molecular Formula Li2O
Molecular Weight (g/mol) 29.88
MDL Number MFCD00016183
InChI Key XZIUWXMPQBQHMC-UHFFFAOYSA-N
IUPAC Name (lithiooxy)lithium
SMILES [Li]O[Li]

Specifications

Melting Point 1427°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.5% (Metal basis)
Solubility Information Soluble in water.
Formula Weight 29.88
Odor Odorless
Sensitivity Air and moisture sensitive
Appearance White powder
Density 2.013
Chemical Name or Material Lithium oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.