missing translation for 'onlineSavingsMsg'
Learn More

Lithium oxalate, 99+%

Codice prodotto. 11356758
Cambia vista
Click to view available options
Quantity:
50 g
250 g
1 kg
Dimensione della confezione:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
11356758 50 g 50g
11346758 250 g 250g
11336758 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11356758 Fornitore Thermo Scientific Alfa Aesar N. del fornitore 013426.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 553-91-3 | C2Li2O4 | 101.90 g/mol

Lithium oxalate is used as lithium battery electrolyte. It is also used as an analytical reagent and reducing reagent. Further, it is used as an intermediate in the manufacture of other chemicals such as pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 553-91-3
Molecular Formula C2Li2O4
Molecular Weight (g/mol) 101.90
MDL Number MFCD00040596
InChI Key YNQRWVCLAIUHHI-UHFFFAOYSA-L
Synonym lithium oxalate, dilithium oxalate, ethanedioic acid, dilithium salt, unii-k737ot0e73, oxalic acid dilithium salt, ethanedioic acid, lithium salt 1:2, oxalic acid dilithium, acmc-1ctwf, c2o4.2li, ksc492a1t
PubChem CID 68383
SMILES [Li+].[Li+].[O-]C(=O)C([O-])=O

Specifica

Melting Point (decomposition)
Density 2.12 g/mL
Quantity 50 g
UN Number UN3288
Merck Index 14,5537
Solubility Information Soluble in water.
IUPAC Name dilithium;oxalate
Formula Weight 101.96
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Lithium oxalate
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Lithium oxalate
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
Indicazioni di pericolo
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Informazioni supplementari
  • MIXTURE LIST-Contain: Lithium oxalate

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.