missing translation for 'onlineSavingsMsg'
Learn More

Lithium oxalate, 99+%

Product Code. 11336758
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336758 1 kg 1kg
11356758 50 g 50g
11346758 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11336758 Supplier Thermo Scientific Alfa Aesar Supplier No. 013426.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 553-91-3 | C2Li2O4 | 101.90 g/mol

Lithium oxalate is used as lithium battery electrolyte. It is also used as an analytical reagent and reducing reagent. Further, it is used as an intermediate in the manufacture of other chemicals such as pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-91-3
Molecular Formula C2Li2O4
Molecular Weight (g/mol) 101.90
MDL Number MFCD00040596
InChI Key YNQRWVCLAIUHHI-UHFFFAOYSA-L
Synonym lithium oxalate, dilithium oxalate, ethanedioic acid, dilithium salt, unii-k737ot0e73, oxalic acid dilithium salt, ethanedioic acid, lithium salt 1:2, oxalic acid dilithium, acmc-1ctwf, c2o4.2li, ksc492a1t
PubChem CID 68383
SMILES [Li+].[Li+].[O-]C(=O)C([O-])=O

Specifications

Melting Point (decomposition)
Density 2.12 g/mL
Quantity 1 kg
UN Number UN3288
Merck Index 14,5537
Solubility Information Soluble in water.
IUPAC Name dilithium(1+) oxalate
Formula Weight 101.96
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Lithium oxalate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.