missing translation for 'onlineSavingsMsg'
Learn More

Lithium octanoate, 99%

Product Code. 11328138
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328138 25 g 25g
11318138 100 g 100g
11338138 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11328138 Supplier Thermo Scientific Alfa Aesar Supplier No. 039371.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16577-52-9 | C8H15LiO2 | 150.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16577-52-9
Molecular Formula C8H15LiO2
Molecular Weight (g/mol) 150.146
MDL Number MFCD00210629
InChI Key BTAUEIDLAAYHSL-UHFFFAOYSA-M
Synonym lithium octanoate, unii-9nq46xlp8b, octanoic acid, lithium salt, 9nq46xlp8b, octanoic acid, lithiumsalt 1:1, lithium caprylate, octanoic acid, lithium salt 1:1, acmc-20akmi, lithium 1+ octanoate, lithium 1+ ion octanoate
PubChem CID 23673717
IUPAC Name lithium;octanoate
SMILES [Li+].CCCCCCCC(=O)[O-]

Specifications

Quantity 25 g
Formula Weight 150.14
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Lithium octanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.