missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium nitride, 99.4% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Lithium nitride is used in metallurgy and chemical synthesis. It is also used to store hydrogen and acts as a source of the nitride ion. It is involved in the preparation of lithium hydride and lithium amide as well as a reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 845°C |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.5% (metals basis) |
Solubility Information | Insoluble in organic solvents. |
IUPAC Name | trilithium;azanide |
Formula Weight | 34.82 |
Sensitivity | Moisture sensitive |
Density | 1.30 g/mL |
Chemical Name or Material | Lithium nitride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'