missing translation for 'onlineSavingsMsg'
Learn More

Lithium Nitrate Ionization Buffer: 1% Li (from carbonate) in 5% HNO3, VHG™

Product Code. 30003111
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003111 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003111 Supplier VHG Labs Supplier No. VHGMLIN1P100
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Lithium Nitrate Ionization Buffer: 1% Li (from carbonate) in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium
CAS 7439-93-2
Methods AA/GFAA
Analyte or Component Names Lithium
Shipping Condition Room Temperature
Molecular Formula Li
Concentration 1%
Synonym Li
InChI Formula InChI=1S/Li
SMILES [Li]
IUPAC Name Lithium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 6.94
Quantity 100 mL
Formula Weight 7.016 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.