Learn More
Lithium nitrate, anhydrous, 99%, Thermo Scientific Chemicals
Description
Used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, and rocket propellant Lithium nitrate is used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, rocket propellant and in solar batteries. It acts as an oxidizing agent in the production of red-colored fireworks and flares; as a catalyst to accelerate the breakdown of nitrogen oxides and as a medium to store heat received from the sun for cooking. It is applied to concrete pavement to withstand weathering effects.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 600°C (decomposition) |
Melting Point | 264°C |
Color | White |
Physical Form | Crystalline |
Quantity | 25 g |
Assay Percent Range | 99% |
Solubility Information | Soluble in water,alcohol,ethanol,methanol,pyridine,ammonia and acetone. |
IUPAC Name | lithium;nitrate |
Formula Weight | 68.95 |
Odor | Odorless |
Show More |
RUO – Research Use Only