missing translation for 'onlineSavingsMsg'
Learn More

Lithium niobium isopropoxide, 99+% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals

Catalog Number 11384858
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
21864-21-1
C18H42LiNbO6
MFCD00210626
This item is not returnable. View return policy
21864-21-1
C18H42LiNbO6
MFCD00210626

Used as solid-state electrolyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Odor Alcohol-like
Linear Formula LiNb(OC3H7)6
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 454.38
Percent Purity ≥99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Lithium niobium isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'