missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium niobium isopropoxide, 99+% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
Description
Used as solid-state electrolyte.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Odor | Alcohol-like |
Linear Formula | LiNb(OC3H7)6 |
Quantity | 100 mL |
UN Number | UN1219 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 454.38 |
Percent Purity | ≥99% |
Physical Form | Liquid |
Assay | (metals basis) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'